missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMEM119 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB103131-100
This item is not returnable.
View return policy
Description
TMEM119 Monoclonal antibody specifically detects TMEM119 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications
| TMEM119 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| OBIF, Osteoblast Induction Factor, TMEM119 | |
| Chinese Hamster Ovary cell line CHO derived human TREM119, Arg26-Met96, Accession # Q4V9L6.1 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Flow Cytometry, Immunohistochemistry | |
| 2699A | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-25 ug/mL | |
| Q4V9L6.1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 338773 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction