missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMEM119 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
242.00 € - 526.00 €
Specifications
| Antigen | TMEM119 |
|---|---|
| Clone | 2699A |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-25 ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18704097
![]() |
R&D Systems
MAB103131-SP |
25 μg |
242.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783607
![]() |
R&D Systems
MAB103131-100 |
100 μg |
526.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM119 Monoclonal antibody specifically detects TMEM119 in Human samples. It is validated for Flow Cytometry, ImmunohistochemistrySpecifications
| TMEM119 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-25 ug/mL | |
| Monoclonal | |
| Rabbit | |
| Human | |
| Q4V9L6.1 | |
| 338773 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2699A | |
| Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| OBIF, Osteoblast Induction Factor, TMEM119 | |
| Chinese Hamster Ovary cell line CHO derived human TREM119, Arg26-Met96, Accession # Q4V9L6.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title