missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMIGD2/CD28H Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB83162-SP
This item is not returnable.
View return policy
Description
TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Specifications
| TMIGD2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2 | |
| Human embryonic kidney cell line HEK-293-derived human TMIGD2/CD28H Met1-Gly150 Accession # Q96BF3 | |
| 25 μg | |
| Primary | |
| Detects human TMIGD2/CD28H in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 953743 | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 50 ug/mL, CyTOF-ready | |
| Q96BF3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 126259 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction