missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TMIGD2/CD28H Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 541.00 €
Specifications
| Antigen | TMIGD2 |
|---|---|
| Clone | 953743 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 50 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725289
![]() |
R&D Systems
MAB83162-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18193451
![]() |
R&D Systems
MAB83162-100 |
100 μg |
541.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMIGD2 Monoclonal specifically detects TMIGD2 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
| TMIGD2 | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 50 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD28H, IGPR1, IGPR-1, MGC23244, transmembrane and immunoglobulin domain containing 2, transmembrane and immunoglobulin domain-containing protein 2 | |
| Human embryonic kidney cell line HEK-293-derived human TMIGD2/CD28H Met1-Gly150 Accession # Q96BF3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TMIGD2/CD28H in direct ELISAs. |
| 953743 | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96BF3 | |
| 126259 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title