missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VE-Cadherin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB938
This item is not returnable.
View return policy
Beschreibung
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human samples. It is validated for Western Blot.
Spezifikation
| VE-Cadherin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1003 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 123433 | |
| Western Blot 1 ug/mL | |
| P33151 | |
| CDH5 | |
| Mouse myeloma cell line NS0-derived recombinant human VE-Cadherin Asp48-Gln593 Accession # P33151 | |
| 500 μg | |
| Primary | |
| Detects human VE-Cadherin in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Cadherin-8, rhCadherin-17, rhE-Cadherin, rhP-Cadherin, or rmVE-Cadherin. | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur