missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VE-Cadherin Antibody, R&D Systems™

Mouse Monoclonal Antibody
208.00 € - 546.00 €
Specifications
| Antigen | VE-Cadherin |
|---|---|
| Clone | 123433 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18775709
![]() |
R&D Systems
MAB938-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18151921
![]() |
R&D Systems
MAB938 |
500 μg |
546.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human samples. It is validated for Western Blot.Specifications
| VE-Cadherin | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin | |
| CDH5 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 123433 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P33151 | |
| 1003 | |
| Mouse myeloma cell line NS0-derived recombinant human VE-Cadherin Asp48-Gln593 Accession # P33151 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VE-Cadherin in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Cadherin-8, rhCadherin-17, rhE-Cadherin, rhP-Cadherin, or rmVE-Cadherin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title