missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VGLUT1/SLC17A7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9054
This item is not returnable.
View return policy
Description
VGLUT1 Monoclonal specifically detects VGLUT1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| VGLUT1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BNPI, BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, SLC17A7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57030 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 732607 | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9P2U7 | |
| SLC17A7 | |
| E.coli-derived recombinant human VGLUT/SLC17A7 Asn93-Thr115 Accession # Q9P2U7 | |
| 100 μg | |
| Primary | |
| Detects human VGLUT/SLC17A7 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction