missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VGLUT1/SLC17A7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
211.00 € - 396.00 €
Specifications
| Antigen | VGLUT1 |
|---|---|
| Clone | 732607 |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18765788
![]() |
R&D Systems
MAB9054-SP |
25 μg |
211.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18746338
![]() |
R&D Systems
MAB9054 |
100 μg |
396.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VGLUT1 Monoclonal specifically detects VGLUT1 in Human samples. It is validated for Immunohistochemistry.Specifications
| VGLUT1 | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BNPI, BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, SLC17A7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1 | |
| SLC17A7 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 732607 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9P2U7 | |
| 57030 | |
| E.coli-derived recombinant human VGLUT/SLC17A7 Asn93-Thr115 Accession # Q9P2U7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VGLUT/SLC17A7 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title