missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VSIG8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9418-SP
This item is not returnable.
View return policy
Description
VSIG8 Monoclonal specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
| VSIG8 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| V-set and immunoglobulin domain containing 8 | |
| Human embryonic kidney cell line HEK293-derived transfected with human VSIG8 Met1-Gly263 Accession # Q5VU13 | |
| 25 μg | |
| Primary | |
| Detects human VSIG8 in direct ELISAs. Stains human VSIG8 transfectants but not irrelevant transfectants in flow cytometry. | |
| Human | |
| IgG1 |
| Flow Cytometry, ELISA | |
| 961823 | |
| Flow Cytometry 0.25 ug/10^6 cells, ELISA | |
| Q5VU13 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 391123 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction