missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VSIG8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
189.00 € - 379.00 €
Specifications
| Antigen | VSIG8 |
|---|---|
| Clone | 961823 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789058
![]() |
R&D Systems
MAB9418-SP |
25 μg |
189.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752959
![]() |
R&D Systems
MAB9418-100 |
100 μg |
379.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VSIG8 Monoclonal specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
| VSIG8 | |
| Flow Cytometry 0.25 ug/10^6 cells, ELISA | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q5VU13 | |
| 391123 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 961823 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| V-set and immunoglobulin domain containing 8 | |
| Human embryonic kidney cell line HEK293-derived transfected with human VSIG8 Met1-Gly263 Accession # Q5VU13 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VSIG8 in direct ELISAs. Stains human VSIG8 transfectants but not irrelevant transfectants in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title