missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyethyl Salicylate, Mikromol™

Product Code. 17954166
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17954166 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17954166 Supplier Mikromol Supplier No. MM0112.000100
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Hydroxyethyl Salicylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salicylic acid-(2-hydroxyethyl) ester
CAS 87-28-5
Analyte or Component Names Salicylic acid-(2-hydroxyethyl) ester
Shipping Condition Room Temperature
Molecular Formula C9 H10 O4
Certifications/Compliance ISO 17034
Synonym Hydroxyethyl Salicylate, Benzoic acid, 2-hydroxy-, 2-hydroxyethyl ester, Salicylic acid, 2-hydroxyethyl ester (6CI,7CI,8CI), Ethylene glycol, monosalicylate (8CI), 2-Hydroxyethyl 2-hydroxybenzoate, 2-Hydroxyethyl salicylate, Espirosal, Ethylene glycol salicylate, GL 7, Glycol monosalicylate, Glycol salicylate, Glysal, Monoglycol salicylate, Norgesic, Rheumacyl, Saliment, Sarocol, Spirosal, beta-Hydroxyethyl salicylate
InChI Formula InChI=1S/C9H10O4/c10-5-6-13-9(12)7-3-1-2-4-8(7)11/h1-4,10-11H,5-6H2
SMILES OCCOC(=O)c1ccccc1O
IUPAC Name 2-hydroxyethyl 2-hydroxybenzoate
Molecular Weight (g/mol) 182.17
Quantity 100 mg
Formula Weight 182.0579 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.