missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyzine.2HCl, Lipomed

Product Code. 30461917
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461917 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461917 Supplier Lipomed Supplier No. LPMHDX1733HC50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Hydroxyzine.2HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxyzine Hydrochloride
CAS 2192-20-3
Analyte or Component Names Hydroxyzine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H27 Cl N2 O2 . 2 Cl H
Synonym Ethanol, 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, hydrochloride (1:2), Ethanol, 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9 CI), 1-(p-Chlorobenzhydryl)-4-[2-(2-hydroxyethoxy)ethyl]diethylenediamine dihydrochloride, Alamon, Atarax, Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI), Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8 CI)
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.Cl.OCCOCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC Name 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]ethanol;dihydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 447.83
Quantity 50 mg
Formula Weight 446.1295 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.