missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyzine Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Hydroxyzine Hydrochloride |
| CAS | 2192-20-3 |
| Analyte or Component Names | Hydroxyzine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 Cl N2 O2 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanol, 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, hydrochloride (1:2), Ethanol, 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9CI), 1-(p-Chlorobenzhydryl)-4-[2-(2-hydroxyethoxy)ethyl]diethylenediamine dihydrochloride, Alamon, Atarax, Atarax P Parenteral soln., Atarax dihydrochloride, Aterax, Atranine A, Durrax, Hydroxizine chloride, Hydroxyzine dihydrochloride, Hydroxyzine hydrochloride, Hydroxyzyne hydrochloride, Neurolax, Orgatrax, QYS, Quiess, Vistaril Parenteral, Vistaril Steraject, Ethanol, 2-[2-[4-(p-chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8CI), 2-[2-[4-(p-Chloro-alpha-phenylbenzyl)-1-piperazinyl]ethoxy]ethanol dihydrochloride |
| InChI Formula | InChI=1S/C21H27ClN2O2.2ClH/c22-20-8-6-19(7-9-20)21(18-4-2-1-3-5-18)24-12-10-23(11-13-24)14-16-26-17-15-25;;/h1-9,21,25H,10-17H2;2*1H |
| Solution Type | Neat |
| SMILES | Cl.Cl.OCCOCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?