missing translation for 'onlineSavingsMsg'
Learn More

Hyoscine Hydrobromide Trihydrate, Mikromol™

Product Code. 18020280
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18020280 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18020280 Supplier Mikromol Supplier No. MM0546.00
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Hyoscine Hydrobromide Trihydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hyoscine Hydrobromide Trihydrate
CAS 6533-68-2
Analyte or Component Names Hyoscine Hydrobromide Trihydrate
Shipping Condition Room Temperature
Molecular Formula C17 H21 N O4 . Br H . 3 H2 O
Certifications/Compliance ISO 17034
Synonym Atropine Imp. F (EP), Hyoscine Butylbromide Imp. A (EP), Hyoscyamine Sulfate Imp. D (EP), Hyoscyamine Imp. D (EP), Hyoscine Hydrobromide Trihydrate, Hyoscyamine Sulphate Imp. D (EP) as Hydrobromide Trihydrate, Hyoscine Butylbromide Imp. A (EP) as Hydrobromide Trihydrate, Atropine Imp. F (EP) as Hydrobromide Trihydrate, (alphaS)-alpha-(Hydroxymethyl)benzeneacetic acid (1alpha,2beta,4beta,5alpha,7beta)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester hydrobromide hydrate (1:1:3), (alphaS)-alpha-(Hydroxymethyl)benzeneacetic acid (1alpha,2beta,4beta,5alpha,7beta)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester hydrobromide trihydrate, alpha-(Hydroxymethyl)benzeneacetic acid [7(S)-(1alpha,2beta,4beta,5alpha,7beta)]-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester hydrobromide trihydrate, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane benzeneacetic acid deriv., (-)-Scopolamine hydrobromide trihydrate, Hyoscine hydrobromide trihydrate, Scopolamine hydrobromide trihydrate, Atropine Impurity F as Hydrobromide Trihydrate, Hyoscine Butylbromide Impurity A as Hydrobromide Trihydrate, Hyoscyamine Sulfate Impurity D, Hyoscyamine Sulfate Impurity D as Hydrobromide Trihydrate, Hyoscyamine Impurity D, Hyoscyamine Impurity D as Hydrobromide Trihydrate
InChI Formula InChI=1S/C17H21NO4.BrH.3H2O/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;;;;/h2-6,11-16,19H,7-9H2,1H3;1H;3*1H2/t11-,12-,13-,14+,15-,16+;;;;/m1..../s1
SMILES O.O.O.Br.CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4
IUPAC Name (αS)-α-(Hydroxymethyl)benzeneacetic acid (1α,2β,4β,5α,7β)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]non-7-yl ester hydrobromide trihydrate
Molecular Weight (g/mol) 438.31
Quantity 500 mg
Formula Weight 437.1049 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.