missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen, Mikromol™

Product Code. 18021520
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18021520 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18021520 Supplier Mikromol Supplier No. MM0002.00
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ibuprofen
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ibuprofen
CAS 15687-27-1
Analyte or Component Names Ibuprofen
Shipping Condition Room Temperature
Molecular Formula C13 H18 O2
Certifications/Compliance ISO 17025
Synonym alpha-Methyl-4-(2-methylpropyl)-benzeneacetic acid, p-Isobutylhydratropic acid, (+/-)-alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid, (+/-)-2-(p-Isobutylphenyl)propionic acid, (+/-)-Ibuprofen, (+/-)-Ibuprophen, (4-Isobutylphenyl)-alpha-methyl-acetic acid, (RS)-Ibuprofen, alpha-(4-Isobutylphenyl)propionic acid, alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid, 2-(4'-Isobutylphenyl)propionic acid, 2-(4-Isobutylphenyl)propanoic acid, 2-(p-Isobutylphenyl)propionic acid, 4-Isobutylhydratropic acid, Act 3, Actiprofen, Adex 200, Adran, Advil, Alaxan, Algi-Flanderil, Algiflex, Algofen, Am-Fam 400, Amibufen, Anafen, Anco, Andran, Anflagen, Antarene, Antiflam, Apo-Ibuprofen, Apsifen, Artofen, Artril, Artril 300, Atril 300, Balkaprofen, Betaprofen, Bloom, Bluton, Brofen, Brufanic, Brufen, Brufen 400, Brufen Retard, Bruflam, Brufort, Buburone, Buluofen, Burana, Butacortelone, Butylenin, Carol, Cobo, Codral Period Pain, Combiflam, Dansida, Dentigoa, Dibufen, Dolgin, Dolgirid, Dolgit, Dolmaral, Dolo-Dolgit, Dolocyl, Dolofen, Dolofen F, Dolomax, Donjust B, Doretrim, Dorival, Drin, Easifon, Ebufac, Emflam, Emflam 200, Emodin, Epobron, Femadon, Fenbid, Fenspan, Focus, Gofen, Gynofug, Haltran, IB 100, IP 82, Ibosure, Ibren, Ibu-Attritin, Ibu-Tab, Ibu-slow, Ibufen, Ibuflamar, Ibufug, Ibugen, Ibugesic, Ibuleve, Ibulgan, Ibumetin, Ibupirac, Ibupril, Ibuprocin, Ibuprofen, Ibuprohm, Ibusal, Ibutad, Ifen, Inabrin, Inflam, Inza, Ipren, Iprogel, Irfen, Isodol, Lamidon, Librofem, Lidifen, Liptan, Lopane, Mensoton, Motrin, Motrin IB, Mynosedin, NSC 256857, Nagifen-D, Napacetin, Nobafon, Nobfelon, Nobgen, Noritis, Norton, Novogent, Novoprofen, Nuprin, Nurofen, Optifen, Opturem, Ostarin, Ostofen, Paduden, Panafen, Pantrop, Paxofen, Pediaprofen, Perofen, Proartinal, Profen, Proflex, Prontalgin, Provon, Quadrax, RD 13621, Rafen, Ranofen, Recidol, Relcofen, Roidenin, Rufin, Rupan, Seclodin, Stelar, Suspren, Syntofene, Tabalon, Tabalon 400, Tarein, Tatanal, Tofen, Trendar, U 18753, Unipron, Upfen, Uprofen, Urem, Zafen, Zofen, dl-Ibuprofen, p-Isobutyl-2-phenylpropionic acid, 4-Isobutyl-a-methylphenylacetic acid
InChI Formula InChI=1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)O
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
Molecular Weight (g/mol) 206.28
Quantity 500 mg
Formula Weight 206.1307 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.