Learn More
ICAM-1 Rabbit, anti-Mouse, Clone: 2E+23, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82827-1-RR-20UL
Description
The ICAM-1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of mouse ICAM-1. This antibody recognizes mouse, rat antigen. The ICAM-1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ICAM-1 | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| CD54, ICAM 1, Icam1, Ly 47, MALA 2 | |
| 20 μL | |
| 15894 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |