missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1 Rabbit, anti-Mouse, Clone: 2E+23, Proteintech
Rabbit Recombinant Monoclonal Antibody
149.00 € - 439.00 €
Specifications
| Antigen | ICAM-1 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18065305
|
Proteintech
82827-1-RR-100UL |
100 μL |
439.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095185
|
Proteintech
82827-1-RR-20UL |
20 μL |
149.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ICAM-1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of mouse ICAM-1. This antibody recognizes mouse, rat antigen. The ICAM-1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ICAM-1 | |
| Western Blot, ELISA | |
| Rabbit | |
| 15894 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 1000 μg/mL | |
| Liquid | |
| Mouse, Rat | |
| CD54, ICAM 1, Icam1, Ly 47, MALA 2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title