missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 1 Antibody (F18), Novus Biologicals™
Rat Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-64387SS
This item is not returnable.
View return policy
Description
IFN-alpha 1 Monoclonal specifically detects IFN-alpha 1 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Functional, Block/Neutralize.
Specifications
| IFN-alpha D | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Blocking/Neutralizing 6.25 x 10^4 neatralizing units/mg | |
| P01572 | |
| IFNA1 | |
| Made to Mouse IFN ALPHA | |
| 0.025 mg | |
| Cancer, Immunology, Innate Immunity | |
| 3439 | |
| Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Functional Assay, Blocking Assay | |
| F18 | |
| Unconjugated | |
| PBS and 0.1% Bovine Serum Albumin with No Preservative | |
| IFL, IFN, IFNA13, IFNAalphaIFN-alpha-1/13, IFN-ALPHA, IFN-alpha 1b, IFN-alphaD, interferon alpha 1b, interferon alpha-1/13, Interferon alpha-D, interferon, alpha 1, LeIF D, MGC138207, MGC138505, MGC138507 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64387 recognizes most isoforms of natural and recombinant murine alpha interferon. It does not cross-react with murine beta or gamma interferon, nor with human alpha interferon. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction