missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha 1 Antibody (F18), Novus Biologicals™
Rat Monoclonal Antibody has been used in 3 publications
294.00 € - 567.00 €
Specifications
| Antigen | IFN-alpha D |
|---|---|
| Clone | F18 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Blocking/Neutralizing 6.25 x 10^4 neatralizing units/mg |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Functional Assay, Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18082776
|
Novus Biologicals
NB100-64387SS |
0.025 mg |
294.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18718123
|
Novus Biologicals
NB100-64387 |
0.1 mg |
567.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-alpha 1 Monoclonal specifically detects IFN-alpha 1 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Functional, Block/Neutralize.Specifications
| IFN-alpha D | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Functional Assay, Blocking Assay | |
| Unconjugated | |
| Rat | |
| Cancer, Immunology, Innate Immunity | |
| P01572 | |
| 3439 | |
| Made to Mouse IFN ALPHA | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| F18 | |
| ELISA, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional, Blocking/Neutralizing 6.25 x 10^4 neatralizing units/mg | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| IFL, IFN, IFNA13, IFNAalphaIFN-alpha-1/13, IFN-ALPHA, IFN-alpha 1b, IFN-alphaD, interferon alpha 1b, interferon alpha-1/13, Interferon alpha-D, interferon, alpha 1, LeIF D, MGC138207, MGC138505, MGC138507 | |
| IFNA1 | |
| IgG1 | |
| Protein G purified | |
| NB100-64387 recognizes most isoforms of natural and recombinant murine alpha interferon. It does not cross-react with murine beta or gamma interferon, nor with human alpha interferon. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title