missing translation for 'onlineSavingsMsg'
Learn More

IFN-gamma Antibody, R&D Systems™

Product Code. 18610199 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18610199 0.1 mg 0.1mg
18607117 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18610199 Supplier Novus Biologicals Supplier No. NBP3113620.1mg
  • ONLINE EXCLUSIVE PRICE
    932.05 €
    793.80 € / 0.1mg
    Save 138.25 €
    14% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

IFN-gamma Polyclonal specifically detects IFN-gamma in Feline samples. It is validated for ELISA∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-gamma
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS
Gene Alias IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Host Species Rabbit
Immunogen Recombinant feline IFN Gamma
Purification Method Affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3458
Target Species Feline
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.