Learn More
IFN-γ Mouse anti-Human, V450, Clone: B27, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560371
Description
The B27 antibody reacts with human interferon-γ (IFN-γ). This is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN-γ has been described. The B27 antibody has been reported not to bind to denatured IFN-γ. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: B27
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IFN-γ Recombinant Protein
Immunocytochemistry, Intracellular Staining
Specifications
| IFN-γ | |
| Monoclonal | |
| Violet Fluor 450 (V450) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| B27 | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Human IFN-γ Recombinant Protein | |
| 120 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.