Learn More
IFN-γ Mouse anti-Human, V450, Clone: B27, BD
Mouse Monoclonal Antibody
183.61 € - 417.30 €
Specifications
| Antigen | IFN-γ |
|---|---|
| Clone | B27 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Violet Fluor 450 (V450) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15810149
|
BD Biosciences
560372 |
30 Tests |
192.00 € 183.61 € / Each Save 8.39 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15800149
|
BD Biosciences
560371 |
120 Tests |
436.00 € 417.30 € / Each Save 18.70 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The B27 antibody reacts with human interferon-γ (IFN-γ). This is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN-γ has been described. The B27 antibody has been reported not to bind to denatured IFN-γ. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: B27
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IFN-γ Recombinant Protein
Immunocytochemistry, Intracellular Staining
Specifications
| IFN-γ | |
| Flow Cytometry | |
| Violet Fluor 450 (V450) | |
| RUO | |
| Human | |
| Human IFN-γ Recombinant Protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| B27 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.