Learn More
Invitrogen™ IFNAR1 Recombinant Rabbit Monoclonal Antibody (120)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560452
 View more versions of this product
Product Code. 30535271
  • 109.00 € / 20µL
Description
IFNAR1 Recombinant Monoclonal Antibody for ELISA
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001, Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFNAR1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P33896
Ifnar1
Recombinant Mouse IFNAR1 protein (Met1-Thr429).
20 μL
Primary
Mouse
Antibody
IgG
ELISA
120
Unconjugated
Ifnar1
alpha-type antiviral protein; AVP; beta-type antiviral protein; CD118; CRF2-1; cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; H3K1; Ifar; IFN R1; IFNalpha/beta R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFNalpha/betaR1; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFNalpha/beta-Ralpha; IFN-alpha-REC; IFNAR; Ifnar1; IFNBR; IFNR1; IFN-R-1; Ifrc; INF-a receptor; Infar; interferon (alpha and beta) receptor 1; interferon (alpha, beta and omega) receptor 1; interferon alpha and beta receptor subunit 1; interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; type I interferon receptor 1
Rabbit
Protein A
RUO
15975
-20°C, Avoid Freeze/Thaw Cycles