missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFNAR1 Recombinant Rabbit Monoclonal Antibody (120)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

109.00 € - 429.00 €

Specifications

Antigen IFNAR1
Clone 120
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30536261
GREENER_CHOICE
View Documents
Invitrogen™
MA560451
100 μL
429.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30535271
View Documents
Invitrogen™
MA560452
20 μL
109.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IFNAR1 Recombinant Monoclonal Antibody for ELISA

Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001, Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).
Specifications

Specifications

IFNAR1
1 mg/mL
ELISA
Unconjugated
Rabbit
RUO
PBS with no preservative
P33896
15975
Recombinant Mouse IFNAR1 protein (Met1-Thr429).
Antibody
120
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Mouse
Ifnar1
alpha-type antiviral protein; AVP; beta-type antiviral protein; CD118; CRF2-1; cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; H3K1; Ifar; IFN R1; IFNalpha/beta R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFNalpha/betaR1; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFNalpha/beta-Ralpha; IFN-alpha-REC; IFNAR; Ifnar1; IFNBR; IFNR1; IFN-R-1; Ifrc; INF-a receptor; Infar; interferon (alpha and beta) receptor 1; interferon (alpha, beta and omega) receptor 1; interferon alpha and beta receptor subunit 1; interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; type I interferon receptor 1
Ifnar1
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.