missing translation for 'onlineSavingsMsg'
Learn More

IGBP1 Antibody (6E3) [Alexa Fluor« 350], Novus Biologicals Biologicals™

Product Code. 30520513 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30520513 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30520513 Supplier Novus Biologicals Supplier No. NBP342985AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGBP1 Monoclonal antibody specifically detects IGBP1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGBP1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6000
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias ALPHA-4, B cell signal transduction molecule alpha 4, bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ), B-cell signal transduction molecule alpha 4, CD79a-binding protein 1, IBP1, immunoglobulin (CD79A) binding protein 1, immunoglobulin-binding protein 1, Protein alpha-4, protein phosphatase 2A, regulatory subunit alpha-4, Renal carcinoma antigen NY-REN-16
Host Species Mouse
Immunogen immunoglobulin(CD79A) binding protein 1 - (Uniprot# P78318)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3476
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.