missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Mouse anti-Human, BB700, Clone: IA6-2, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566539
This item is not returnable.
View return policy
Description
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Flow Cytometry.
Specifications
| IgD | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| 25 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ |
| Flow Cytometry | |
| IA6-2 | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction