missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgE Antibody (7H2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11624-0.1mg
This item is not returnable.
View return policy
Description
IgE Monoclonal antibody specifically detects IgE in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| IgE | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Equine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 7H2 | |
| Western Blot, ELISA 1:40 - 1:400, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| IgE, immunoglobulin heavy constant epsilon | |
| Equine IgE | |
| 0.1 mg | |
| Adaptive Immunity, Immunology | |
| 3497 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction