missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgE Antibody (7H2), Novus Biologicals™
Mouse Monoclonal Antibody
190.00 € - 463.00 €
Specifications
| Antigen | IgE |
|---|---|
| Clone | 7H2 |
| Dilution | Western Blot, ELISA 1:40 - 1:400, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637698
|
Novus Biologicals
NBP3-11624-0.1mg |
0.1 mg |
463.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667727
|
Novus Biologicals
NBP3-11624-0.025mg |
0.025 mg |
190.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgE Monoclonal antibody specifically detects IgE in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| IgE | |
| Western Blot, ELISA 1:40 - 1:400, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Purified | |
| RUO | |
| Equine | |
| IgE, immunoglobulin heavy constant epsilon | |
| Equine IgE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 7H2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology | |
| PBS | |
| 3497 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title