missing translation for 'onlineSavingsMsg'
Learn More

IGFBP-2 Antibody (CL13563) [HRP], Novus Biologicals™

Product Code. 30524693 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30524693 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30524693 Supplier Novus Biologicals Supplier No. NBP343841H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IGFBP-2 Monoclonal antibody specifically detects IGFBP-2 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP-2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL13563
Conjugate HRP
Formulation PBS
Gene Alias BP2, IBP2, IBP-2, IGF-binding protein 2, IGFBP-2, IGF-BP53, insulin-like growth factor binding protein 2 (36kD), insulin-like growth factor binding protein 2, 36kDa, insulin-like growth factor-binding protein 2
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P18065, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytokines and Growth Factors, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3485
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.