missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-3 Antibody (MM0342-6U23) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12364-0.025MG
This item is not returnable.
View return policy
Description
IGFBP-3 Monoclonal specifically detects IGFBP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IGFBP-3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-2000, Immunohistochemistry 1:20-200, Immunohistochemistry-Paraffin 1:20-200 | |
| CAA46087 | |
| IGFBP3 | |
| Human IGFBP3 | |
| 0.025 mg | |
| Apoptosis, Prostate Cancer | |
| 3486 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| MM0342-6U23 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction