missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP-3 Antibody (MM0342-6U23) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
195.00 € - 507.00 €
Specifications
| Antigen | IGFBP-3 |
|---|---|
| Clone | MM0342-6U23 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18073527
|
Novus Biologicals
NBP2-12364-0.025MG |
0.025 mg |
195.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18799703
|
Novus Biologicals
NBP2-12364 |
||||||||
Description
IGFBP-3 Monoclonal specifically detects IGFBP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IGFBP-3 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3 | |
| IGFBP3 | |
| Protein G purified |
| MM0342-6U23 | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Prostate Cancer | |
| CAA46087 | |
| 3486 | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title