missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG (Immunoglobulin Gamma Heavy Chain) (B-Cell Marker) Mouse, Clone: rIG266, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3500-MSM10-P1
Additional Details : Weight : 0.01000kg
Specifications
IgG (Immunoglobulin Gamma Heavy Chain) (B-Cell Marker) | |
Cytoplasm, Cell Surface and Secreted | |
rIG266 | |
Unconjugated | |
Mouse | |
75kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Purified human Ig Gamma Chain | |
293T, Raji or hPBL cells. Tonsil or Spleen. | |
100 μg | |
Primary | |
IgG1 |