missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Rabbit, Clone: IG1707R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 3500-RBM9-P1
This item is not returnable.
View return policy
Specifications
| IgG | |
| Cytoplasm, Cell Surface and Secreted | |
| IG1707R | |
| Unconjugated | |
| IGHG | |
| Purified human Ig Gamma Chain | |
| 293T, Raji or hPBL cells. Tonsil or Spleen. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| Rabbit | |
| 75kDa | |
| Protein A/G Purified | |
| RUO | |
| 3500 | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction