missing translation for 'onlineSavingsMsg'
Learn More

IgG Rabbit, Clone: IG1707R, enQuireBio™

Rabbit Monoclonal Antibody

199.00 € - 478.00 €

Specifications

Antigen IgG
Clone IG1707R
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15910228
View Documents
EnquireBio
3500-RBM9-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15920228
View Documents
EnquireBio
3500-RBM9-P1
100 μg
ONLINE EXCLUSIVE PRICE
554.40 €
478.00 € / 100µg
Save 76.40 €
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

IgG
200μg/mL
Monoclonal
Rabbit
10mM PBS with or without 0.05% BSA and 0.05% azide.
IGHG
IgG κ
Protein A/G Purified
Cytoplasm, Cell Surface and Secreted
293T, Raji or hPBL cells. Tonsil or Spleen.
IG1707R
Immunohistochemistry
Unconjugated
RUO
3500
Purified human Ig Gamma Chain
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
75kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.