missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IkB-beta, p Ser23 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54761
This item is not returnable.
View return policy
Description
IkB-beta p Ser23 Polyclonal antibody specifically detects IkB-beta p Ser23 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| IkB-beta (p Ser23) | |
| Polyclonal | |
| Unconjugated | |
| Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| NFKBIB | |
| IkB-beta (Phospho-Ser23) antibody was raised against a peptide sequence around phosphorylation site of serine 23 (L-G-S (p) -L-G) derived from Human IkappaB-b | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody detects endogenous level of IkB-beta; only when phosphorylated at serine 23. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| Immunohistochemistry | |
| 1 mg/mL | |
| Immunohistochemistry 1:50-1:100 | |
| beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta | |
| Rabbit | |
| 48 kDa | |
| 0.1 mg | |
| Immunology, Innate Immunity, Phospho Specific | |
| 4793 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction