missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IkB-beta, p Ser23 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00 € - 688.00 €
Specifications
| Concentration | 1 mg/mL |
|---|---|
| Dilution | Immunohistochemistry 1:50-1:100 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18290510
|
Novus Biologicals
NBP2-54761 |
0.1 mg |
688.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18239504
|
Novus Biologicals
NBP2-54761-0.05MG |
0.05 mg |
441.00 €
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685246
|
Novus Biologicals
NBP2-54761-0.025mg |
0.025 mg |
195.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IkB-beta Polyclonal specifically detects IkB-beta in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Human, Mouse, Rat | |
| 4793 | |
| IkB-beta (Phospho-Ser23) antibody was raised against a peptide sequence around phosphorylation site of serine 23 (L-G-S (p) -L-G) derived from Human IkappaB-b | |
| Primary |
| Immunohistochemistry 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta | |
| NFKBIB | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title