missing translation for 'onlineSavingsMsg'
Learn More

IKBKAP Antibody [mFluor Violet 500 SE], Novus Biologicals Biologicals™

Product Code. 30492747 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30492747 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30492747 Supplier Novus Biologicals Supplier No. NBP335099MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IKBKAP Polyclonal antibody specifically detects IKBKAP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKBKAP
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1093-1332 of human IKBKAP (NP_003631.2).,, Sequence:, LRLVYKYNRLDIIETNVKPSILEAQKNYMAFLDSQTATFSRHKKRLLVVRELKEQAQQAGLDDEVPHGQESDLFSETSSVVSGSEMSGKYSHSNSRISARSSKNRRKAERKKHSLKEGSPLEDLALLEALSEVVQNTENLKDEVYHILKVLFLFEFDEQGRELQKAFEDTLQLMERSLPEIWTLTYQQNSATPVLGPNSTANSIMASYQQQKTSVPVLDAELFIPPKINRRTQWKLSLLD
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8518
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.