missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
310.00 € - 758.00 €
Specifications
| Antigen | IL-13 |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677147
|
Novus Biologicals
NBP3-11366-0.025mg |
0.025 mg |
310.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18651439
|
Novus Biologicals
NBP3-11366-0.1mg |
0.1 mg |
758.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-13 Polyclonal antibody specifically detects IL-13 in Canine samples. It is validated for Western Blot, ELISASpecifications
| IL-13 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Canine | |
| ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 | |
| Recombinant canine IL-13 | |
| Primary | |
| Store at 4°C. Do not freeze. |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| PBS | |
| 3596 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title