missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17RE Antibody (46N7E3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-27368SS
This item is not returnable.
View return policy
Description
IL-17RE Monoclonal specifically detects IL-17RE in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-17RE | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 5-10ug/ml∼, Flow Cytometry reported in scientific literature (PMID 27534549), Immunohistochemistry, Immunohistochemistry-Paraffin 10ug/ml∼ | |
| IL-17 receptor E, interleukin 17 receptor E | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 132014 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 46N7E3 | |
| Unconjugated | |
| Q8NFR9 | |
| IL17RE | |
| amino acids (97-199) MVHLLVQkSkkSSTFkFYRRHkMPAPAQRkLLPRRHLSEkSHHISIPSPDISHkGLRSkR∼TQPSDPETWESLPRLDSQRHGGPEFSFDLLPEARAIRVTISSGPE | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido