missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18/IL-1F4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
214.00 € - 522.00 €
Specifications
| Antigen | IL-18/IL-1F4 |
|---|---|
| Clone | 1072F |
| Dilution | Neutralization 0.05-0.3 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18796469
![]() |
R&D Systems
MAB9124-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762418
![]() |
R&D Systems
MAB9124 |
100 μg |
522.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Neutralization.Specifications
| IL-18/IL-1F4 | |
| Neutralization 0.05-0.3 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
| IL18 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1072F | |
| Neutralization | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q14116 | |
| 3606 | |
| E. coli-derived recombinant human IL-18/IL-1F4 Tyr37-Asp197 Accession # Q14116 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-18/IL-1F4 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title