Learn More
IL-2 Rat anti-Human, FITC, Clone: MQ1-17H12, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554565
Description
The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2). The immunogen used to generate the MQ1-17H12 hybridoma was recombinant human IL-2. This is a neutralizing antibody.
Host Species: Rat
Clone: MQ1-17H12
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Recombinant Human IL-2 protein
Intracellular Staining
Specifications
| IL-2 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human IL-2 protein | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| MQ1-17H12 | |
| FITC | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.