Learn More
IL-2 Rat anti-Human, FITC, Clone: MQ1-17H12, BD
Rat Monoclonal Antibody
524.11 €
Specifications
| Antigen | IL-2 |
|---|---|
| Clone | MQ1-17H12 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
The MQ1-17H12 monoclonal antibody specifically binds to the multifunctional cytokine, human Interleukin-2 (IL-2). IL-2 is produced by activated T cells and has multiple functions that can affect the growth, proliferation, differentiation and survival of many different target cell types including T cells, B cells, NK cells, monocytes and macrophages. The immunogen used to generate the MQ1-17H12 hybridoma was purified recombinant human IL-2 protein. The MQ1-17H12 antibody reportedly neutralizes the biological activity of human IL-2.
Specifications
| IL-2 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Human | |
| IgG2a κ | |
| Affinity Purified |
| MQ1-17H12 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.