missing translation for 'onlineSavingsMsg'
Learn More

IL-33 Antibody (40015C), Allophycocyanin, R&D Systems™

Product Code. 18605753 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18605753 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18605753 Supplier Novus Biologicals Supplier No. FAB36253A
  • ONLINE EXCLUSIVE PRICE
    773.47 €
    657.30 € / 0.1mL
    Save 116.17 €
    15% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Goat Monoclonal Antibody

IL-33 Monoclonal specifically detects IL-33 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-33
Applications ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 40015C
Conjugate APC
Dilution Western Blot, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry
Formulation PBS
Gene Alias C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2
Host Species Goat
Immunogen E.coli-derived recombinant human IL-33, Ser112-Thr270, Accession # O95760
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cellular Markers, Core ESC Like Genes, Immunology, Inflammation, Innate Immunity, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 90865
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.