missing translation for 'onlineSavingsMsg'
Learn More

IL-7 Antibody (CL11132) [Janelia Fluor« 669], Novus Biologicals Biologicals™

Product Code. 30529127 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30529127 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30529127 Supplier Novus Biologicals Supplier No. NBP344211JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-7 Monoclonal antibody specifically detects IL-7 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-7
Applications Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL11132
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias IL-7interleukin-7, interleukin 7
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P13232, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3574
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.