missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7 Antibody (CL11132), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07998-25ul
This item is not returnable.
View return policy
Description
IL-7 Monoclonal antibody specifically detects IL-7 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| IL-7 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| CL11132 | |
| Immunocytochemistry/ Immunofluorescence 2-10 μg/ml | |
| IL-7interleukin-7, interleukin 7 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: GRKPAALGEAQPTKSLEE | |
| 25 μg | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cell Markers | |
| 3574 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction