missing translation for 'onlineSavingsMsg'
Learn More

IL-7 Antibody (CL11132), Novus Biologicals™

Mouse Monoclonal Antibody

366.00 € - 625.00 €

Specifications

Antigen IL-7
Clone CL11132
Dilution Immunocytochemistry/ Immunofluorescence 2-10 μg/ml
Applications Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18638507
View Documents
Novus Biologicals
NBP3-07998-25ul
25 μg
366.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18693489
View Documents
Novus Biologicals
NBP3-07998-100ul
100 μg
625.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-7 Monoclonal antibody specifically detects IL-7 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

IL-7
Immunocytochemistry/ Immunofluorescence 2-10 μg/ml
Monoclonal
Purified
RUO
Human
IL-7interleukin-7, interleukin 7
This antibody was developed against a synthetic peptide corresponding to amino acids: GRKPAALGEAQPTKSLEE
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
CL11132
Immunofluorescence
Unconjugated
Mouse
Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cell Markers
PBS, pH 7.2, containing 40% glycerol
3574
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.