Learn More
IL6R Antibody (B-N12), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-14598UV
397.80 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18533157
  • 470.00 € / 0.10mL
Description
IL6R Monoclonal antibody specifically detects IL6R in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL6R
Monoclonal
DyLight 350
CD126, CD126 antigen, gp80, IL-6 receptor subunit alpha, IL-6R 1, IL-6R subunit alpha, IL-6R-1, IL6RA, IL-6RA, IL-6R-alpha, interleukin 6 receptor, interleukin 6 receptor alpha subunit, interleukin-6 receptor subunit alpha, Membrane glycoprotein 80, MGC104991
gp80 transfected CHO cell line
0.1 mL
Cancer, Cytokine Research, Immunology, Innate Immunity
3570
Store at 4°C in the dark.
IgG1 κ
Flow Cytometry, ELISA
B-N12
50mM Sodium Borate
Mouse
Affinity purified
RUO
Primary
Human
Purified