missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ILDR2 Antibody (bapotulimab) - IgG2SA, Novus Biologicals™
Shop All Bio Techne ProductsDescription
ILDR2 Monoclonal antibody specifically detects ILDR2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | ILDR2 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | bapotulimab |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| Gene Accession No. | Q71H61 |
| Gene Alias | immunoglobulin-like domain containing receptor 2 |
| Host Species | Human |
| Immunogen | ILDR2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?