missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILK Antibody (3A9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37448-0.025ml
This item is not returnable.
View return policy
Description
ILK Monoclonal specifically detects ILK in Human, Mouse, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ILK | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| ILK | |
| Purified recombinant fragment of human ILK (AA: 97-244) expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate, Monkey | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3A9 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| 59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK | |
| Mouse | |
| 51.4 kDa | |
| 0.025 mL | |
| Protein Kinase, Signal Transduction | |
| 3611 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction