missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILK Antibody (3A9), Novus Biologicals™
Mouse Monoclonal Antibody
203.00 € - 552.00 €
Specifications
| Antigen | ILK |
|---|---|
| Clone | 3A9 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673226
|
Novus Biologicals
NBP2-37448-0.025ml |
0.025 mL |
203.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18140718
|
Novus Biologicals
NBP2-37448 |
0.1 mL |
552.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILK Monoclonal specifically detects ILK in Human, Mouse, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ILK | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Primate, Monkey | |
| 59 kDa serine/threonine-protein kinase, DKFZp686F1765, EC 2.7.11.1, ILK1, ILK-1, ILK2, ILK-2, integrin-linked kinase, integrin-linked kinase-2, integrin-linked protein kinase, P59, p59ILK | |
| ILK | |
| IgG1 | |
| Unpurified |
| 3A9 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Protein Kinase, Signal Transduction | |
| Ascites with 0.03% Sodium Azide | |
| 3611 | |
| Purified recombinant fragment of human ILK (AA: 97-244) expressed in E. Coli. | |
| Primary | |
| 51.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title